accessibility menu, dialog, popup

UserName

H3COHOHO

CAS RN 106-14-9

IUPAC Name: 12-hydroxyoctadecanoic acid
Synonyms: DL-12-hydroxy stearic acid
Molecular Weight (g/mol): 300.48
Molecular Formula: C18H36O3
InChi Key: ULQISTXYYBZJSJ-UHFFFAOYNA-N
SMILES: CCCCCCC(O)CCCCCCCCCCC(O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)