missing translation for 'onlineSavingsMsg'
Learn More

12-Hydroxystearic Acid 80.0+%, TCI America™

Supplier:  TCI America H0308500G

 View more versions of this product

Catalog No. H0308500G

Only null left

Description

Description

Specifications

Specifications

12-Hydroxystearic Acid
78°C
180°C
MFCD00004592
12-hydroxystearic acid, octadecanoic acid, 12-hydroxy, harwax a, cerit fac 3, hydrofol acid 200, dl-12-hydroxystearic acid, ceroxin gl, barolub fto, loxiol g 21, stearic acid, 12-hydroxy
CCCCCCC(CCCCCCCCCCC(=O)O)O
300.483
CHEBI:85208
≥80.0% (GC)
106-14-9
White-Yellow
C18H36O3
500 g
ULQISTXYYBZJSJ-UHFFFAOYSA-N
12-hydroxyoctadecanoic acid
7789
300.48
Crystalline Powder

Chemical Identifiers

106-14-9
300.483
ULQISTXYYBZJSJ-UHFFFAOYSA-N
7789
12-hydroxyoctadecanoic acid
C18H36O3
MFCD00004592
12-hydroxystearic acid, octadecanoic acid, 12-hydroxy, harwax a, cerit fac 3, hydrofol acid 200, dl-12-hydroxystearic acid, ceroxin gl, barolub fto, loxiol g 21, stearic acid, 12-hydroxy
CHEBI:85208
CCCCCCC(CCCCCCCCCCC(=O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1340

RTECSNumber : WI3850000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions