missing translation for 'onlineSavingsMsg'
Learn More

12-Hydroxystearic Acid 80.0+%, TCI America™

Catalog No. H030825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H030825G 25 g
H0308500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H030825G Supplier TCI America Supplier No. H030825G

Chemical Identifiers

CAS 106-14-9
Molecular Formula C18H36O3
Molecular Weight (g/mol) 300.483
MDL Number MFCD00004592
InChI Key ULQISTXYYBZJSJ-UHFFFAOYSA-N
Synonym 12-hydroxystearic acid, octadecanoic acid, 12-hydroxy, harwax a, cerit fac 3, hydrofol acid 200, dl-12-hydroxystearic acid, ceroxin gl, barolub fto, loxiol g 21, stearic acid, 12-hydroxy
PubChem CID 7789
ChEBI CHEBI:85208
IUPAC Name 12-hydroxyoctadecanoic acid
SMILES CCCCCCC(CCCCCCCCCCC(=O)O)O

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 180°C
Quantity 25 g
Formula Weight 300.48
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 12-Hydroxystearic Acid
EINECSNumber (2)-1340
RTECSNumber WI3850000
TSCA Yes