missing translation for 'onlineSavingsMsg'
Learn More

12-Amino-1-dodecanol 98.0+%, TCI America™

Catalog No. A15241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15241G 1 g
A15245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15241G Supplier TCI America Supplier No. A15241G

Chemical Identifiers

CAS 67107-87-3
Molecular Formula C12H27NO
Molecular Weight (g/mol) 201.354
MDL Number MFCD01851111
InChI Key IIWXYWWVCBRBCJ-UHFFFAOYSA-N
PubChem CID 5182020
IUPAC Name 12-aminododecan-1-ol
SMILES C(CCCCCCO)CCCCCN

Specifications

Melting Point 81°C
Color White
Quantity 1 g
Formula Weight 201.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 12-Amino-1-dodecanol