accessibility menu, dialog, popup

UserName

HONH2

CAS RN 67107-87-3

IUPAC Name: 12-aminododecan-1-ol
Synonyms: 12-aminododecan-1-ol
Molecular Weight (g/mol): 201.35
Molecular Formula: C12H27NO
InChi Key: IIWXYWWVCBRBCJ-UHFFFAOYSA-N
SMILES: NCCCCCCCCCCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results