missing translation for 'onlineSavingsMsg'
Learn More

11H-Benzo[b]fluoren-11-one 98.0+%, TCI America™

Catalog No. B39325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B39325G Supplier TCI America Supplier No. B39325G
Only null left

Chemical Identifiers

CAS 3074-03-1
Molecular Formula C17H10O
Molecular Weight (g/mol) 230.27
MDL Number MFCD00215960
InChI Key MLMNDNOSVOKYMT-UHFFFAOYSA-N
PubChem CID 18311
IUPAC Name benzo[b]fluoren-11-one
SMILES C1=CC=C2C=C3C(=CC2=C1)C4=CC=CC=C4C3=O

Specifications

Melting Point 155°C
Color Yellow
Quantity 5 g
Formula Weight 230.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 11H-Benzo[b]fluoren-11-one
RTECSNumber DF6407000
TSCA No