missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11H-Benzo[a]carbazole 98.0+%, TCI America™
$287.06 - $831.37
Chemical Identifiers
| CAS | 239-01-0 |
|---|---|
| Molecular Formula | C16H11N |
| Molecular Weight (g/mol) | 217.271 |
| MDL Number | MFCD00215939 |
| InChI Key | MYKQKWIPLZEVOW-UHFFFAOYSA-N |
| Synonym | 1,2-Benzocarbazole |
| PubChem CID | 9196 |
| IUPAC Name | 11H-benzo[a]carbazole |
| SMILES | C1=CC=C2C(=C1)C=CC3=C2NC4=CC=CC=C34 |
Chemical Identifiers
| 239-01-0 | |
| 217.271 | |
| MYKQKWIPLZEVOW-UHFFFAOYSA-N | |
| 9196 | |
| C1=CC=C2C(=C1)C=CC3=C2NC4=CC=CC=C34 |
| C16H11N | |
| MFCD00215939 | |
| 1,2-Benzocarbazole | |
| 11H-benzo[a]carbazole |
Specifications
| 239-01-0 | |
| White-Yellow | |
| MFCD00215939 | |
| 1,2-Benzocarbazole | |
| C1=CC=C2C(=C1)C=CC3=C2NC4=CC=CC=C34 | |
| 217.271 | |
| 217.27 | |
| Crystalline Powder |
| 233°C | |
| C16H11N | |
| 1 g | |
| MYKQKWIPLZEVOW-UHFFFAOYSA-N | |
| 11H-benzo[a]carbazole | |
| 9196 | |
| ≥98.0% (GC) | |
| 11H-Benzo[a]carbazole |
Safety and Handling
RTECSNumber : DE7025000
TSCA : No