accessibility menu, dialog, popup

UserName

CAS RN 239-01-0

NH

CAS RN 239-01-0

IUPAC Name: 11H-benzo[a]carbazole
Synonyms: 11H-benzo[a]carbazole
Molecular Weight (g/mol): 217.27
Molecular Formula: C16H11N
InChi Key: MYKQKWIPLZEVOW-UHFFFAOYSA-N
SMILES: N1C2=C(C=CC=C2)C2=C1C1=CC=CC=C1C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results