missing translation for 'onlineSavingsMsg'
Learn More

(11bS)-2,6-Bis[bis[3,5-bis(trifluoromethyl)phenyl]hydroxymethyl]-3,5-dihydrospiro[4H-dinaphth[2,1-c:1',2'-e] 97+%, TCI America™

Catalog No. B397050MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B397050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B397050MG Supplier TCI America Supplier No. B397050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1197922-04-5
Molecular Formula C60H36BrF24NO3
Molecular Weight (g/mol) 1354.818
InChI Key UVHZWWMJSAGSJH-UHFFFAOYSA-M
PubChem CID 44597114
SMILES C1COCC[N+]12CC3=C(C4=CC=CC=C4C=C3C(C5=CC(=CC(=C5)C(F)(F)F)C(F)(F)F)(C6=CC(=CC(=C6)C(F)(F)F)C(F)(F)F)O)C7=C(C2)C(=CC8=CC=CC=C87)C(C9=CC(=CC(=C9)C(F)(F)F)C(F)(F)F)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)O.[Br-]

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 1354.82
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (11bS)-2,6-Bis[bis[3,5-bis(trifluoromethyl)phenyl]hydroxymethyl]-3,5-dihydrospiro[4H-dinaphth[2,1-c: