missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HEPES (Ultra Pure)

Catalog No. 11344041
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11344041 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11344041 Supplier Gibco™ Supplier No. 11344041

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A zwitterionic organic chemical buffering agent commonly used in cell culture media

  • Formulated using 11g per liter of water
  • Addition of 10 to 25mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator
  • Buffering choice for many cell culture systems because it is membrane impermeable
  • Has limited effect on biochemical reactions
  • Chemically and enzymatically stable
  • Low visible and UV light absorbance

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Form Powder
Product Type HEPES Buffer
pH 6.8 to 8.2
Quantity 1 kg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.