missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Keto-β-boswellic acid, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 17019-92-0 |
| Physical Form | Neat |
| Linear Formula | C30H46O4 |
| MDL Number | MFCD06656312 |
| Synonym | 11-Oxo-beta-boswellic acid; 3 alpha-Hydroxy-11-oxours-12-en-24-oic acid; KBA |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 470.68 |
| Percent Purity | ≥95.0% (HPLC) |
| Show More |