missing translation for 'onlineSavingsMsg'
Learn More

11-Chloro-1-undecene, 97%

Catalog No. p-7035723
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53449AA 5 mL
AAH53449AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53449AA Supplier Thermo Scientific Chemicals Supplier No. H53449AA
Only null left

CAS: 872-17-3 | C11H21Cl | 188.74 g/mol

11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 872-17-3
Molecular Formula C11H21Cl
Molecular Weight (g/mol) 188.74
MDL Number MFCD00671354
InChI Key PSEVKFKRYVAODC-UHFFFAOYSA-N
Synonym 11-chloro-1-undecene, 1-undecene, 11-chloro, 10-undecenyl chloride, undecylenyl chloride, acmc-1bjnc, undec-10-enyl chloride, 11-chloro-1-undecene #
PubChem CID 543805
IUPAC Name 11-chloroundec-1-ene
SMILES ClCCCCCCCCCC=C

Specifications

Density 0.995
Boiling Point 108°C to 109°C (6 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.451
Linear Formula Cl(CH2)9CH=CH2
Quantity 5 mL
Solubility Information Soluble in water.
Formula Weight 188.74
Percent Purity 97%
Chemical Name or Material 11-Chloro-1-undecene
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only