accessibility menu, dialog, popup

UserName

CAS RN 872-17-3

IUPAC Name: 11-chloroundec-1-ene
Synonyms: 11-chloroundec-1-ene
Molecular Weight (g/mol): 188.74
Molecular Formula: C11H21Cl
InChi Key: PSEVKFKRYVAODC-UHFFFAOYSA-N
SMILES: ClCCCCCCCCCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)