missing translation for 'onlineSavingsMsg'
Learn More

11-Azidoundecyltrimethoxysilane, 94%

Catalog No. p-7032723
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34298MD 250 mg
AAH3429803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34298MD Supplier Thermo Scientific Chemicals Supplier No. H34298MD
Only null left

CAS: 334521-23-2 | C14H31N3O3Si | 317.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 334521-23-2
Molecular Formula C14H31N3O3Si
Molecular Weight (g/mol) 317.51
MDL Number MFCD11982878
InChI Key ACSJXAKDVNDMME-UHFFFAOYSA-N
Synonym 11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide
PubChem CID 46779110
SMILES CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC

Specifications

Boiling Point 100°C to 105°C (0.04mmHg)
Quantity 250 mg
Linear Formula N3(CH2)11Si(OCH3)3
IUPAC Name (11-azidoundecyl)trimethoxysilane
Formula Weight 317.51
Percent Purity 94%
Sensitivity Moisture sensitive
Chemical Name or Material 11-Azidoundecyltrimethoxysilane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only