accessibility menu, dialog, popup

UserName

CAS RN 334521-23-2

IUPAC Name: (11-azidoundecyl)trimethoxysilane
Synonyms: (11-azidoundecyl)trimethoxysilane
Molecular Weight (g/mol): 317.51
Molecular Formula: C14H31N3O3Si
InChi Key: ACSJXAKDVNDMME-UHFFFAOYSA-N
SMILES: CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results