missing translation for 'onlineSavingsMsg'
Learn More

11-Azidoundecyltriethoxysilane, 95%

Catalog No. AAH33524MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33524MD 250 mg
AAH3352403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33524MD Supplier Thermo Scientific Chemicals Supplier No. H33524MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 663171-33-3 | C17H37N3O3Si | 359.586 g/mol

This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 663171-33-3
Molecular Formula C17H37N3O3Si
Molecular Weight (g/mol) 359.586
MDL Number MFCD11982879
InChI Key WCDFLYKXACDAES-UHFFFAOYSA-N
Synonym azidoundecyltriethoxysilane, 11-azidoundecyltriethoxysilane, 11-azidoundecyl triethoxysilane
PubChem CID 46779111
IUPAC Name 11-azidoundecyl(triethoxy)silane
SMILES CCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC

Specifications

Boiling Point 110°C to 115°C (0.04mmHg)
Quantity 250 mg
Linear Formula N3(CH2)11Si(OCH2CH3)3
Formula Weight 359.58
Percent Purity 95%
Refractive Index 1.4390-1.4430
Sensitivity Moisture sensitive
Chemical Name or Material 11-Azidoundecyltriethoxysilane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only