missing translation for 'onlineSavingsMsg'
Learn More

(11-Azidoundecyl)trimethoxysilane 93.0+%, TCI America™

Catalog No. A2783100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2783100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2783100MG Supplier TCI America Supplier No. A2783100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 334521-23-2
Molecular Formula C14H31N3O3Si
Molecular Weight (g/mol) 317.51
MDL Number MFCD11982878
InChI Key ACSJXAKDVNDMME-UHFFFAOYSA-N
Synonym 11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide
PubChem CID 46779110
IUPAC Name (11-azidoundecyl)trimethoxysilane
SMILES CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC

Specifications

Color Yellow
Boiling Point 105°C
Quantity 100 mg
Formula Weight 317.51
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material (11-Azidoundecyl)trimethoxysilane