missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Azido-3,6,9-trioxaundecan-1-amine 95.0+%, TCI America™
Supplier: TCI America A23631G
Specifications
| 11-Azido-3,6,9-trioxaundecan-1-amine | |
| C8H18N4O3 | |
| 1 g | |
| 1-amino-11-azido-3,6,9-trioxaundecane, azido-peg3-amine, 11-azido-3,6,9-trioxaundecan-1-amine, 2-2-2-2-azidoethoxy ethoxy ethoxy ethanamine, 1-2-2-aminoethoxy ethoxy-2-2-azidoethoxy ethane, 11-azido-3 6 9-trioxaundecan-1-amine, ethanamine, 2-2-2-2-azidoethoxy ethoxy ethoxy, 2-2-2-2-azidoethoxy ethoxy ethoxy ethylamine | |
| C(COCCOCCOCCN=[N+]=[N-])N | |
| 218.257 | |
| 218.26 | |
| Liquid |
| 134179-38-7 | |
| MFCD00269874 | |
| 2735 | |
| FPVCVHVTMPCZTH-UHFFFAOYSA-N | |
| 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethanamine | |
| 2735280 | |
| ≥95.0% (T) |
Chemical Identifiers
| 134179-38-7 | |
| 218.257 | |
| FPVCVHVTMPCZTH-UHFFFAOYSA-N | |
| 2735280 | |
| C(COCCOCCOCCN=[N+]=[N-])N |
| C8H18N4O3 | |
| MFCD00269874 | |
| 1-amino-11-azido-3,6,9-trioxaundecane, azido-peg3-amine, 11-azido-3,6,9-trioxaundecan-1-amine, 2-2-2-2-azidoethoxy ethoxy ethoxy ethanamine, 1-2-2-aminoethoxy ethoxy-2-2-azidoethoxy ethane, 11-azido-3 6 9-trioxaundecan-1-amine, ethanamine, 2-2-2-2-azidoethoxy ethoxy ethoxy, 2-2-2-2-azidoethoxy ethoxy ethoxy ethylamine | |
| 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethanamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator