accessibility menu, dialog, popup

UserName

CAS RN 134179-38-7

IUPAC Name: 1-[2-(2-aminoethoxy)ethoxy]-2-(2-azidoethoxy)ethane
Synonyms: 1-[2-(2-aminoethoxy)ethoxy]-2-(2-azidoethoxy)ethane
Molecular Weight (g/mol): 218.26
Molecular Formula: C8H18N4O3
InChi Key: FPVCVHVTMPCZTH-UHFFFAOYSA-N
SMILES: NCCOCCOCCOCCN=[N+]=[N-]
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)