missing translation for 'onlineSavingsMsg'
Learn More

11-Aminoundecyltriethoxysilane, 90+%

Catalog No. p-7032395
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33835MD 250 mg
AAH3383503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33835MD Supplier Thermo Scientific Chemicals Supplier No. H33835MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 116821-45-5 | C17H39NO3Si | 333.588 g/mol

Used as primary and secondary intermediate for pharmaceutical and chemical additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical and chemical additives.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.

Chemical Identifiers

CAS 116821-45-5
Molecular Formula C17H39NO3Si
Molecular Weight (g/mol) 333.588
MDL Number MFCD08272792
InChI Key LPWZCJFZJCOBHO-UHFFFAOYSA-N
PubChem CID 18944844
IUPAC Name 11-triethoxysilylundecan-1-amine
SMILES CCO[Si](CCCCCCCCCCCN)(OCC)OCC

Specifications

Boiling Point 110°C to 115°C (0.02mmHg)
Quantity 250 mg
Sensitivity Moisture sensitive
Linear Formula H2N(CH2)11Si(OCH2CH3)3
Synonym 11-triethoxysilyl-1-undecanamine, 11-aminoundecyltriethoxysilane, 11-triethoxysilyl undecan-1-amine, 1-undecanamine,11-triethoxysilyl, 11-aminoundecyl triethoxysilane, acmc-20mmty, 1-undecanamine, 11-triethoxysilyl
Formula Weight 333.59
Percent Purity 95%
Chemical Name or Material 11-Aminoundecyltriethoxysilane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only