accessibility menu, dialog, popup

UserName

CAS RN 116821-45-5

IUPAC Name: (11-aminoundecyl)triethoxysilane
Synonyms: (11-aminoundecyl)triethoxysilane
Molecular Weight (g/mol): 333.59
Molecular Formula: C17H39NO3Si
InChi Key: LPWZCJFZJCOBHO-UHFFFAOYSA-N
SMILES: CCO[Si](CCCCCCCCCCCN)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results