missing translation for 'onlineSavingsMsg'
Learn More

11-Aminoundecanoic acid, 97%

Catalog No. AC104712500
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104712500 250 g
AC104710050 5 g
AC104711000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104712500 Supplier Thermo Scientific Chemicals Supplier No. 104712500

CAS: 2432-99-7 | C11H23NO2 | 201.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2432-99-7
Molecular Formula C11H23NO2
Molecular Weight (g/mol) 201.31
MDL Number MFCD00008150
InChI Key GUOSQNAUYHMCRU-UHFFFAOYSA-N
Synonym undecanoic acid, 11-amino, 11-aminoundecylic acid, 11-amino-undecanoic acid, omega-aminoundecanoic acid, ccris 39, unii-sfu34976hb, rilsan t, aminoundecanoicacid, reilsan es natural, 11-amino-undecanoicaci
PubChem CID 17083
ChEBI CHEBI:82387
IUPAC Name 11-aminoundecanoic acid
SMILES C(CCCCCN)CCCCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 187°C to 190°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula H2N(CH2)10CO2H
Quantity 250 g
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in butanol
Formula Weight 201.31
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 11-Aminoundecanoic acid, 99%
Show More Show Less
EINECSNumber 219-417-6
RTECSNumber YQ2293000
TSCA TSCA

RUO – Research Use Only