CAS RN 2432-99-7
IUPAC Name:
11-aminoundecanoic acid
Synonyms:
11-aminoundecanoic acid
Molecular Weight (g/mol):
201.31
Molecular Formula:
C11H23NO2
InChi Key:
GUOSQNAUYHMCRU-UHFFFAOYSA-N
SMILES:
NCCCCCCCCCCC(O)=O
1
–
1
of
1
results
1
Promotions Available
11-Aminoundecanoic acid, 97%
11-Aminoundecanoic acid, 97%
CAS: 2432-99-7 Molecular Formula: C11H23NO2 Molecular Weight (g/mol): 201.31 MDL Number: MFCD00008150 InChI Key: GUOSQNAUYHMCRU-UHFFFAOYSA-N Synonym: undecanoic acid, 11-amino,11-aminoundecylic acid,11-amino-undecanoic acid,omega-aminoundecanoic acid,ccris 39,unii-sfu34976hb,rilsan t,aminoundecanoicacid,reilsan es natural,11-amino-undecanoicaci PubChem CID: 17083 ChEBI: CHEBI:82387 IUPAC Name: 11-aminoundecanoic acid SMILES: C(CCCCCN)CCCCC(=O)O