missing translation for 'onlineSavingsMsg'
Learn More

11-[4-(4-Butylphenylazo)phenoxy]undecyl Methacrylate 97.0+%, TCI America™

Catalog No. m146325g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M146325G 25 g
M14635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M146325G Supplier TCI America Supplier No. M146325G

Chemical Identifiers

CAS 942230-11-7
Molecular Formula C31H44N2O3
Molecular Weight (g/mol) 492.704
MDL Number MFCD06797121
InChI Key YKWVMKBZDQOJNN-UHFFFAOYSA-N
Synonym Methacrylic Acid 11-[4-(4-Butylphenylazo)phenoxy]undecyl Ester
PubChem CID 44630394
IUPAC Name 11-[4-[(4-butylphenyl)diazenyl]phenoxy]undecyl 2-methylprop-2-enoate
SMILES CCCCC1=CC=C(C=C1)N=NC2=CC=C(C=C2)OCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Melting Point 76°C
Quantity 25 g
Formula Weight 492.70
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 11-[4-(4-Butylphenylazo)phenoxy]undecyl Methacrylate