Learn More
Thermo Scientific™ SduI, Ppu21I Buffer (10X)
Optimize the performance of SduI and Ppu21I restriction enzymes with this buffer, premixed with BSA for enhanced stability.
Supplier: Thermo Scientific™ B23
Description
To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.
restriction enzymes exhibit 100% of their certified activity in the recommended buffer.
The following enzymes require unique buffers for optimal digestion:Â
- AarI
- AjiI
- BamHI
- BfuI
- Bpu10I
- BseXI
- Bsp143I
- Cfr9I
- Cfr10I
- Eam1105I
- Ecl136II
- Eco52I
- EcoRI
- KpnI
- Lsp1109I
- PacI
- PasI
- Ppu21I
- SacI
- ScaI
- SdaI
- SduI
- SgeI
- TaqI
Specifications
| 1 mL |
For Research Use Only. Not for use in diagnostic procedures.