missing translation for 'onlineSavingsMsg'
Learn More

1000 μg/mL 8270 MegaMix Standard (76 Components) In Methylene Chloride (3-methylphenol and 4-methylphenol at 500 μg/mL), Restek

Catalog No. 06611944
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06611944 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06611944 Supplier Chromatographic Supplier No. RK31850

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fewest mixtures needed for calibration and matrix spikes

  • Mixtures formulated for maximum stability
  • Contains most routinely analyzed compounds

Notes:

  • 1,2-diphenylhydrazine (8270-listed analyte) decomposes to azobenzene (mix component) in injector
  • N-nitrosodiphenylamine (8270-listed analyte) decomposes to diphenylamine (mix component) in injector

Specifications

Solution Contains 76 components
Shipping Condition Ambient
Concentration 1000 μg/mL
Technique Calibration and Matrix Spikes
Certifications/Compliance Certified reference material
Solvent or Matrix Methylene Chloride
Quantity 1 mL