missing translation for 'onlineSavingsMsg'
Learn More

10-Undecynoic Acid 98.0+%, TCI America™

Catalog No. U00541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00541G 1 g
U00545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00541G Supplier TCI America Supplier No. U00541G

Chemical Identifiers

CAS 2777-65-3
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD00014389
InChI Key OAOUTNMJEFWJPO-UHFFFAOYSA-N
Synonym 10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference
PubChem CID 31039
IUPAC Name undec-10-ynoic acid
SMILES C#CCCCCCCCCC(=O)O

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 180°C
Quantity 1 g
Formula Weight 182.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10-Undecynoic Acid
RTECSNumber YQ3683000
TSCA No