missing translation for 'onlineSavingsMsg'
Learn More

10-Undecynoic acid, 96%

CAS: 2777-65-3 | C11H18O2 | 182.263 g/mol

Supplier:  Thermo Scientific Chemicals A1381514

 View more versions of this product

Catalog No. AAA1381514


Only null left

Description

Description

10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

Solubility
Sparingly Soluble in water.

Notes
Store at 4°C. Store away from incompatible materials such as oxidizing agents
Specifications

Specifications

10-Undecynoic acid
40°C to 43°C
161°C (321°F)
C11H18O2
25 g
10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference
OAOUTNMJEFWJPO-UHFFFAOYSA-N
undec-10-ynoic acid
31039
96%
2777-65-3
174°C to 176°C (4 mmHg)
Odorless
MFCD00014389
1704918
Sparingly Soluble in water.
C#CCCCCCCCCC(=O)O
182.263
182.26

Chemical Identifiers

2777-65-3
182.263
OAOUTNMJEFWJPO-UHFFFAOYSA-N
31039
C#CCCCCCCCCC(=O)O
C11H18O2
MFCD00014389
10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference
undec-10-ynoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-471-8

RTECSNumber : YQ3683000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only