missing translation for 'onlineSavingsMsg'
Learn More

10-Undecynoic acid, 96%

Catalog No. AAA1381506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1381506 5 g
AAA1381514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1381506 Supplier Thermo Scientific Chemicals Supplier No. A1381506

CAS: 2777-65-3 | C11H18O2 | 182.263 g/mol

10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

Solubility
Sparingly Soluble in water.

Notes
Store at 4°C. Store away from incompatible materials such as oxidizing agents

Chemical Identifiers

CAS 2777-65-3
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD00014389
InChI Key OAOUTNMJEFWJPO-UHFFFAOYSA-N
Synonym 10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference
PubChem CID 31039
IUPAC Name undec-10-ynoic acid
SMILES C#CCCCCCCCCC(=O)O

Specifications

Melting Point 40°C to 43°C
Boiling Point 174°C to 176°C (4 mmHg)
Flash Point 161°C (321°F)
Odor Odorless
Quantity 5 g
Beilstein 1704918
Solubility Information Sparingly Soluble in water.
Formula Weight 182.26
Percent Purity 96%
Chemical Name or Material 10-Undecynoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-471-8
RTECSNumber YQ3683000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only