missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoyl Chloride 98.0+%, TCI America™

Supplier:  TCI America U0008250ML

 View more versions of this product

Catalog No. U0008250ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10-Undecenoyl Chloride
Yellow
C11H19ClO
250 mL
10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
ClC(=O)CCCCCCCCC=C
202.72
202.72
Liquid
38460-95-6
93°C
MFCD00000772
3265
MZFGYVZYLMNXGL-UHFFFAOYSA-N
undec-10-enoyl chloride
38042
≥98.0% (T)

Chemical Identifiers

38460-95-6
202.72
MZFGYVZYLMNXGL-UHFFFAOYSA-N
38042
ClC(=O)CCCCCCCCC=C
C11H19ClO
MFCD00000772
10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
undec-10-enoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YQ2991000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions