missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoyl Chloride 98.0+%, TCI America™

Catalog No. p-7137837
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U000825ML 25 mL
U0008250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U000825ML Supplier TCI America Supplier No. U000825ML

Chemical Identifiers

CAS 38460-95-6
Molecular Formula C11H19ClO
Molecular Weight (g/mol) 202.72
MDL Number MFCD00000772
InChI Key MZFGYVZYLMNXGL-UHFFFAOYSA-N
Synonym 10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
PubChem CID 38042
IUPAC Name undec-10-enoyl chloride
SMILES ClC(=O)CCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 93°C
Quantity 25 mL
UN Number 3265
Formula Weight 202.72
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 10-Undecenoyl Chloride
RTECSNumber YQ2991000
TSCA Yes