missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Undecenoic Acid 98.0+%, TCI America™
$32.28 - $72.15
Chemical Identifiers
| CAS | 112-38-9 |
|---|---|
| Molecular Formula | C11H20O2 |
| Molecular Weight (g/mol) | 184.279 |
| MDL Number | MFCD00004442 |
| InChI Key | FRPZMMHWLSIFAZ-UHFFFAOYSA-N |
| Synonym | 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid |
| PubChem CID | 5634 |
| ChEBI | CHEBI:35045 |
| IUPAC Name | undec-10-enoic acid |
| SMILES | C=CCCCCCCCCC(=O)O |
Chemical Identifiers
| C11H20O2 | |
| MFCD00004442 | |
| 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid | |
| CHEBI:35045 | |
| C=CCCCCCCCCC(=O)O |
Specifications
| 112-38-9 | |
| 275°C | |
| MFCD00004442 | |
| FRPZMMHWLSIFAZ-UHFFFAOYSA-N | |
| undec-10-enoic acid | |
| 5634 | |
| 184.28 | |
| Crystals |
| White-Yellow | |
| C11H20O2 | |
| 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid | |
| C=CCCCCCCCCC(=O)O | |
| 184.279 | |
| CHEBI:35045 | |
| ≥98.0% (GC,T) | |
| 10-Undecenoic Acid |
Safety and Handling
EINECSNumber : (2)-0609
RTECSNumber : YQ2975000
TSCA : Yes