Learn More
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%
CAS: 944721-52-2 | C20H29F5O5Si | 472.524 g/mol
$99.22 - $280.68
Chemical Identifiers
| CAS | 944721-52-2 |
|---|---|
| Molecular Formula | C20H29F5O5Si |
| Molecular Weight (g/mol) | 472.524 |
| MDL Number | MFCD13181915 |
| InChI Key | KSYPNQIBGIOYPZ-UHFFFAOYSA-N |
| Synonym | 11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate |
| PubChem CID | 23582157 |
| IUPAC Name | (2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate |
| SMILES | CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH34117MD
|
Thermo Scientific Chemicals
H34117MD |
250 mg |
Each for $99.22
|
|
|||||
|
AAH3411703
|
Thermo Scientific Chemicals
H3411703 |
1 g |
Each for $280.68
|
|
|||||
Description
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.
Chemical Identifiers
| 944721-52-2 | |
| 472.524 | |
| KSYPNQIBGIOYPZ-UHFFFAOYSA-N | |
| 23582157 | |
| CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC |
| C20H29F5O5Si | |
| MFCD13181915 | |
| 11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate | |
| (2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate |
Specifications
| 145°C to 150°C (0.05mmHg) | |
| 250 mg | |
| MFCD13181915 | |
| KSYPNQIBGIOYPZ-UHFFFAOYSA-N | |
| (2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate | |
| 23582157 | |
| 95% | |
| Moisture sensitive |
| 944721-52-2 | |
| C20H29F5O5Si | |
| 11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate | |
| CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC | |
| 472.524 | |
| 472.52 | |
| 1.4370-1.4410 | |
| 10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only