missing translation for 'onlineSavingsMsg'
Learn More

10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%

CAS: 944721-52-2 | C20H29F5O5Si | 472.524 g/mol

Supplier:  Thermo Scientific Chemicals H3411703

 View more versions of this product

Catalog No. AAH3411703


Only null left
Explore available promotions

Description

Description

It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.
Specifications

Specifications

145°C to 150°C (0.05mmHg)
944721-52-2
C20H29F5O5Si
11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate
CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC
472.524
472.52
1.4370-1.4410
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane
1 g
MFCD13181915
KSYPNQIBGIOYPZ-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate
23582157
95%
Moisture sensitive

Chemical Identifiers

944721-52-2
472.524
KSYPNQIBGIOYPZ-UHFFFAOYSA-N
23582157
CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC
C20H29F5O5Si
MFCD13181915
11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate
(2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only