missing translation for 'onlineSavingsMsg'
Learn More

10-Methyl-9-phenylacridinium Perchlorate 98.0+%, TCI America™

Catalog No. M17751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17751G 1 g
M17755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17751G Supplier TCI America Supplier No. M17751G

Chemical Identifiers

CAS 36519-61-6
Molecular Formula C20H16ClNO4
Molecular Weight (g/mol) 369.801
InChI Key NRNCXGCGIKATGE-UHFFFAOYSA-M
PubChem CID 11326154
IUPAC Name 10-methyl-9-phenylacridin-10-ium;perchlorate
SMILES C[N+]1=C2C=CC=CC2=C(C3=CC=CC=C31)C4=CC=CC=C4.[O-]Cl(=O)(=O)=O

Specifications

Quantity 1 g
UN Number 1479
Formula Weight 369.80
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 10-Methyl-9-phenylacridinium Perchlorate