missing translation for 'onlineSavingsMsg'
Learn More

10-Methyl-9(10H)-acridone 98.0+%, TCI America™

Supplier:  TCI America M079225G

 View more versions of this product

Catalog No. M079225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10-Methyl-9(10H)-acridone
203°C
MFCD00005024
n-methylacridone, 10-methylacridin-9 10h-one, 10-methyl-9 10h-acridone, 10-methyl-9-acridanone, 9 10h-acridinone, 10-methyl, 10-methylacridone, 9-acridanone, 10-methyl, n-methyl-9-acridone, 10-methylacridon, n-methyl-acridone
CN1C2=CC=CC=C2C(=O)C3=CC=CC=C31
209.248
209.25
Crystalline Powder
719-54-0
C14H11NO
25 g
XUVKSPPGPPFPQN-UHFFFAOYSA-N
10-methylacridin-9-one
69751
≥98.0% (N)

Chemical Identifiers

719-54-0
209.248
XUVKSPPGPPFPQN-UHFFFAOYSA-N
69751
CN1C2=CC=CC=C2C(=O)C3=CC=CC=C31
C14H11NO
MFCD00005024
n-methylacridone, 10-methylacridin-9 10h-one, 10-methyl-9 10h-acridone, 10-methyl-9-acridanone, 9 10h-acridinone, 10-methyl, 10-methylacridone, 9-acridanone, 10-methyl, n-methyl-9-acridone, 10-methylacridon, n-methyl-acridone
10-methylacridin-9-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions