missing translation for 'onlineSavingsMsg'
Learn More

10-Methyl-9(10H)-acridone 98.0+%, TCI America™

Catalog No. p-7132363
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07925G 5 g
M079225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07925G Supplier TCI America Supplier No. M07925G

Chemical Identifiers

CAS 719-54-0
Molecular Formula C14H11NO
Molecular Weight (g/mol) 209.248
MDL Number MFCD00005024
InChI Key XUVKSPPGPPFPQN-UHFFFAOYSA-N
Synonym n-methylacridone, 10-methylacridin-9 10h-one, 10-methyl-9 10h-acridone, 10-methyl-9-acridanone, 9 10h-acridinone, 10-methyl, 10-methylacridone, 9-acridanone, 10-methyl, n-methyl-9-acridone, 10-methylacridon, n-methyl-acridone
PubChem CID 69751
IUPAC Name 10-methylacridin-9-one
SMILES CN1C2=CC=CC=C2C(=O)C3=CC=CC=C31

Specifications

Melting Point 203°C
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 10-Methyl-9(10H)-acridone