missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-10-Camphorsulfonic Acid 98.0+%, TCI America™
$64.56 - $539.21
Chemical Identifiers
| CAS | 3144-16-9 |
|---|---|
| Molecular Formula | C10H15O4S |
| Molecular Weight (g/mol) | 231.29 |
| MDL Number | MFCD00064157,MFCD00074827 |
| InChI Key | MIOPJNTWMNEORI-XVKPBYJWSA-M |
| Synonym | 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid |
| PubChem CID | 65617 |
| ChEBI | CHEBI:55403 |
| IUPAC Name | [(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate |
| SMILES | CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2 |
Chemical Identifiers
| 3144-16-9 | |
| 231.29 | |
| MIOPJNTWMNEORI-XVKPBYJWSA-M | |
| 65617 | |
| [(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate |
| C10H15O4S | |
| MFCD00064157,MFCD00074827 | |
| 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid | |
| CHEBI:55403 | |
| CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2 |
Specifications
| 3144-16-9 | |
| White-Yellow | |
| MFCD00064157,MFCD00074827 | |
| 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid | |
| CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2 | |
| 231.29 | |
| CHEBI:55403 | |
| ≥98.0% (T) | |
| (+)-10-Camphorsulfonic Acid |
| 200°C | |
| C10H15O4S | |
| 3261 | |
| MIOPJNTWMNEORI-XVKPBYJWSA-M | |
| [(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate | |
| 65617 | |
| 232.29 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0309
RTECSNumber : ED1550000
TSCA : Yes