missing translation for 'onlineSavingsMsg'
Learn More

(+)-10-Camphorsulfonic Acid 98.0+%, TCI America™

Catalog No. C001525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C001525G 25 g
C0015100G 100 g
C0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C001525G Supplier TCI America Supplier No. C001525G

Chemical Identifiers

CAS 3144-16-9
Molecular Formula C10H15O4S
Molecular Weight (g/mol) 231.29
MDL Number MFCD00064157,MFCD00074827
InChI Key MIOPJNTWMNEORI-XVKPBYJWSA-M
Synonym 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
PubChem CID 65617
ChEBI CHEBI:55403
IUPAC Name [(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate
SMILES CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 232.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-10-Camphorsulfonic Acid
EINECSNumber (4)-0309
RTECSNumber ED1550000
TSCA Yes