missing translation for 'onlineSavingsMsg'
Learn More

(+)-10-Camphorsulfonic Acid 98.0+%, TCI America™

Supplier:  TCI America C0015500G

 View more versions of this product

Catalog No. C0015500G


Only null left

Description

Description

Specifications

Specifications

(+)-10-Camphorsulfonic Acid
200°C
C10H15O4S
500 g
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
231.29
CHEBI:55403
≥98.0% (T)
3144-16-9
White-Yellow
MFCD00064157,MFCD00074827
3261
MIOPJNTWMNEORI-XVKPBYJWSA-M
[(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate
65617
232.29
Crystalline Powder

Chemical Identifiers

3144-16-9
231.29
MIOPJNTWMNEORI-XVKPBYJWSA-M
65617
[(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate
C10H15O4S
MFCD00064157,MFCD00074827
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
CHEBI:55403
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0309

RTECSNumber : ED1550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions