missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-10-Camphorsulfonic Acid 98.0+%, TCI America™
Supplier: TCI America C0972100G
Specifications
| (-)-10-Camphorsulfonic Acid | |
| 195°C | |
| C10H16O4S | |
| 100 g | |
| l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid | |
| CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2 | |
| 232.29 | |
| CHEBI:55401 | |
| ≥98.0% (T) |
| 35963-20-3 | |
| White-Yellow | |
| MFCD00150753,MFCD00064158 | |
| 3261 | |
| MIOPJNTWMNEORI-MHPPCMCBSA-N | |
| [(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid | |
| 5771688 | |
| 232.29 | |
| Crystalline Powder |
Chemical Identifiers
| 35963-20-3 | |
| 232.29 | |
| MIOPJNTWMNEORI-MHPPCMCBSA-N | |
| 5771688 | |
| [(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid |
| C10H16O4S | |
| MFCD00150753,MFCD00064158 | |
| l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid | |
| CHEBI:55401 | |
| CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2 |
Safety and Handling
EINECSNumber : (4)-0309
TSCA : No