missing translation for 'onlineSavingsMsg'
Learn More

(-)-10-Camphorsulfonic Acid 98.0+%, TCI America™

Catalog No. p-7124716
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C097225G 25 g
C0972100G 100 g
C0972500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C097225G Supplier TCI America Supplier No. C097225G

Chemical Identifiers

CAS 35963-20-3
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00150753,MFCD00064158
InChI Key MIOPJNTWMNEORI-MHPPCMCBSA-N
Synonym l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
PubChem CID 5771688
ChEBI CHEBI:55401
IUPAC Name [(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
SMILES CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 232.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-10-Camphorsulfonic Acid
EINECSNumber (4)-0309
TSCA No