missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Bromodecanoic Acid 97.0+%, TCI America™
$77.84 - $244.92
Chemical Identifiers
| CAS | 50530-12-6 |
|---|---|
| Molecular Formula | C10H19BrO2 |
| Molecular Weight (g/mol) | 251.164 |
| MDL Number | MFCD00014388 |
| InChI Key | PGVRSPIEZYGOAD-UHFFFAOYSA-N |
| Synonym | 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid |
| PubChem CID | 142712 |
| IUPAC Name | 10-bromodecanoic acid |
| SMILES | C(CCCCC(=O)O)CCCCBr |
Chemical Identifiers
| 50530-12-6 | |
| 251.164 | |
| PGVRSPIEZYGOAD-UHFFFAOYSA-N | |
| 142712 | |
| C(CCCCC(=O)O)CCCCBr |
| C10H19BrO2 | |
| MFCD00014388 | |
| 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid | |
| 10-bromodecanoic acid |
Specifications
| 50530-12-6 | |
| C10H19BrO2 | |
| 5 g | |
| PGVRSPIEZYGOAD-UHFFFAOYSA-N | |
| 10-bromodecanoic acid | |
| 142712 | |
| ≥97.0% (GC) | |
| 10-Bromodecanoic Acid |
| 42°C | |
| MFCD00014388 | |
| 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid | |
| C(CCCCC(=O)O)CCCCBr | |
| 251.164 | |
| 251.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No