missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Bromodecanoic Acid 97.0+%, TCI America™
Supplier: TCI America B226425G
Specifications
| 10-Bromodecanoic Acid | |
| 42°C | |
| MFCD00014388 | |
| 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid | |
| C(CCCCC(=O)O)CCCCBr | |
| 251.164 | |
| 251.16 | |
| Crystalline Powder |
| 50530-12-6 | |
| C10H19BrO2 | |
| 25 g | |
| PGVRSPIEZYGOAD-UHFFFAOYSA-N | |
| 10-bromodecanoic acid | |
| 142712 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 50530-12-6 | |
| 251.164 | |
| PGVRSPIEZYGOAD-UHFFFAOYSA-N | |
| 142712 | |
| C(CCCCC(=O)O)CCCCBr |
| C10H19BrO2 | |
| MFCD00014388 | |
| 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid | |
| 10-bromodecanoic acid |
Safety and Handling
TSCA : No