missing translation for 'onlineSavingsMsg'
Learn More

10-Bromo-10'-(2-naphthyl)-9,9'-bianthracene 98.0+%, TCI America™

Catalog No. B4896200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4896200MG 200 mg
B48961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4896200MG Supplier TCI America Supplier No. B4896200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1172087-81-8
Molecular Formula C38H23Br
Molecular Weight (g/mol) 559.506
MDL Number MFCD22987563
InChI Key OUMDUWOVDLPNDY-UHFFFAOYSA-N
PubChem CID 91972118
IUPAC Name 9-bromo-10-(10-naphthalen-2-ylanthracen-9-yl)anthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=C7C=CC=CC7=C(C8=CC=CC=C86)Br

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 559.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 10-Bromo-10′-(2-naphthyl)-9,9′-bianthracene