accessibility menu, dialog, popup

UserName

CAS RN 1172087-81-8

IUPAC Name: 10-bromo-10'-(naphthalen-2-yl)-9,9'-bianthracene
Synonyms: 10-bromo-10'-(naphthalen-2-yl)-9,9'-bianthracene
Molecular Weight (g/mol): 559.51
Molecular Formula: C38H23Br
InChi Key: OUMDUWOVDLPNDY-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=C(C2=CC3=CC=CC=C3C=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results