missing translation for 'onlineSavingsMsg'
Learn More

10-Bromo-1-decene 95.0+%, TCI America™

Catalog No. B28491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28491G 1 g
B28495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28491G Supplier TCI America Supplier No. B28491G

Chemical Identifiers

CAS 62871-09-4
Molecular Formula C10H19Br
Molecular Weight (g/mol) 219.166
InChI Key JVVPJOMYWVYPOF-UHFFFAOYSA-N
Synonym 9-Decenyl Bromide
PubChem CID 543384
IUPAC Name 10-bromodec-1-ene
SMILES C=CCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 219.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 10-Bromo-1-decene
EINECSNumber (2)-0067
TSCA No