missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Bromo-1-decanol 85.0+%, TCI America™
$182.89 - $558.30
Chemical Identifiers
| CAS | 53463-68-6 |
|---|---|
| Molecular Formula | C10H21BrO |
| Molecular Weight (g/mol) | 237.18 |
| MDL Number | MFCD00041681 |
| InChI Key | LGZMUUBPTDRQQM-UHFFFAOYSA-N |
| Synonym | 10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p |
| PubChem CID | 104507 |
| IUPAC Name | 10-bromodecan-1-ol |
| SMILES | OCCCCCCCCCCBr |
Chemical Identifiers
| 53463-68-6 | |
| 237.18 | |
| LGZMUUBPTDRQQM-UHFFFAOYSA-N | |
| 104507 | |
| OCCCCCCCCCCBr |
| C10H21BrO | |
| MFCD00041681 | |
| 10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p | |
| 10-bromodecan-1-ol |
Specifications
| 53463-68-6 | |
| 169°C | |
| MFCD00041681 | |
| 10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p | |
| OCCCCCCCCCCBr | |
| 237.18 | |
| 237.18 | |
| Liquid |
| Yellow | |
| C10H21BrO | |
| 5 g | |
| LGZMUUBPTDRQQM-UHFFFAOYSA-N | |
| 10-bromodecan-1-ol | |
| 104507 | |
| ≥85.0% (GC) | |
| 10-Bromo-1-decanol |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer