Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
10-Bromo-1-decanol, 95%
Description
10-Bromo-1-decanol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
10-Bromo-1-decanol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It is miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
10-Bromo-1-decanol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It is miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications
Specifications
| Density | 1.19 |
| Boiling Point | 165°C to 169°C (10 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.476 |
| Quantity | 1 g |
| Beilstein | 1738593 |
| Solubility Information | It is miscible with water. |
| Formula Weight | 237.19 |
| Percent Purity | 95% |
| Chemical Name or Material | 10-Bromo-1-decanol |
Safety and Handling
| Hazard Category | H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 258-572-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only