missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Amino-1-decanol 98.0+%, TCI America™
$290.79 - $993.54
Chemical Identifiers
| CAS | 23160-46-5 |
|---|---|
| Molecular Formula | C10H23NO |
| Molecular Weight (g/mol) | 173.3 |
| MDL Number | MFCD02094313 |
| InChI Key | SWVSKCPPMNGBGL-UHFFFAOYSA-N |
| PubChem CID | 4465355 |
| IUPAC Name | 10-aminodecan-1-ol |
| SMILES | C(CCCCCO)CCCCN |
Chemical Identifiers
| 23160-46-5 | |
| 173.3 | |
| SWVSKCPPMNGBGL-UHFFFAOYSA-N | |
| 10-aminodecan-1-ol |
| C10H23NO | |
| MFCD02094313 | |
| 4465355 | |
| C(CCCCCO)CCCCN |
Specifications
| 23160-46-5 | |
| White-Yellow | |
| C10H23NO | |
| 1 g | |
| C(CCCCCO)CCCCN | |
| 173.3 | |
| 173.30 | |
| Crystalline Powder |
| 72°C | |
| 154°C | |
| MFCD02094313 | |
| SWVSKCPPMNGBGL-UHFFFAOYSA-N | |
| 10-aminodecan-1-ol | |
| 4465355 | |
| ≥98.0% (GC,T) | |
| 10-Amino-1-decanol |
Safety and Handling
TSCA : No